missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-methylcyclohexane 98.0+%, TCI America™

Catalog No. E04221ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04221ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04221ML Supplier TCI America Supplier No. E04221ML

Chemical Identifiers

CAS 3728-54-9
Molecular Formula C9H18
Molecular Weight (g/mol) 126.24
MDL Number MFCD00067158
InChI Key XARGIVYWQPXRTC-UHFFFAOYNA-N
PubChem CID 35413
IUPAC Name 1-ethyl-2-methylcyclohexane
SMILES CCC1CCCCC1C

Specifications

Color Colorless
Boiling Point 156°C
Quantity 1 mL
UN Number 3295
Formula Weight 126.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2-methylcyclohexane