missing translation for 'onlineSavingsMsg'
Learn More

(1-Ethoxyethylidene)malononitrile, 99+%, Thermo Scientific Chemicals

Catalog No. AC170291000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC170291000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC170291000 Supplier Thermo Scientific Chemicals Supplier No. 170291000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5417-82-3
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00001852
InChI Key BOSVWXDDFBSSIZ-UHFFFAOYSA-N
Synonym 1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene malononitrile, 2-1-ethoxyethylidene propanedinitrile, propanedinitrile, 1-ethoxyethylidene, 1-ethoxyethylidenemalononitrile, 1-ethoxyethylidene propanedinitrile, 2-1-ethoxyethylidene-propanedinitrile, propanedinitrile, 2-1-ethoxyethylidene, ethoxyethylidene methane-1,1-dicarbonitrile, malononitrile, 1-ethoxyethylidene-6ci,7ci,8ci
PubChem CID 79450
IUPAC Name 2-(1-ethoxyethylidene)propanedinitrile
SMILES CCOC(=C(C#N)C#N)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(OC2H5)=C(CN)2
Quantity 100 g
Formula Weight 136.15
Percent Purity 99+%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material (1-Ethoxyethylidene)malononitrile, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENT
EINECSNumber 226-521-5
RTECSNumber TY1510000
TSCA TSCA

RUO – Research Use Only