missing translation for 'onlineSavingsMsg'
Learn More

1-(Ethoxycarbonyl)-4-piperidinecarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7129392
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12081G 1 g
E12085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12081G Supplier TCI America Supplier No. E12081G

Chemical Identifiers

CAS 118133-15-6
Molecular Formula C9H15NO4
Molecular Weight (g/mol) 201.222
MDL Number MFCD07339354
InChI Key BVEBMPFJSMEGJX-UHFFFAOYSA-N
Synonym 1-(Ethoxycarbonyl)isonipecotic Acid
PubChem CID 6618886
IUPAC Name 1-ethoxycarbonylpiperidine-4-carboxylic acid
SMILES CCOC(=O)N1CCC(CC1)C(=O)O

Specifications

Melting Point 69°C
Color White-Yellow
Quantity 1 g
Formula Weight 201.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Ethoxycarbonyl)-4-piperidinecarboxylic Acid