missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 95%

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol

Supplier:  Thermo Scientific Chemicals 259050025

 View more versions of this product

Catalog No. AC259050025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Ethoxy-2-propanol
-100°C
0.8970g/mL
42°C
94% min. Remark: balance 2-ethoxy-1-propanol (GC)
C5H12O2
C2H5OCH2CH(OH)CH3
2.5 L
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
1-ethoxypropan-2-ol
15287
104.14
Liquid
1569-02-4
Colorless
132°C
Authentic
Glass Bottle
1.4050 to 1.409
MFCD00067050
0.897
Solubility in water: soluble
CCOCC(C)O
104.15
2 mPa.s (20°C)
95%

Chemical Identifiers

1569-02-4
104.15
JOLQKTGDSGKSKJ-UHFFFAOYNA-N
15287
CCOCC(C)O
C5H12O2
MFCD00067050
1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
1-ethoxypropan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Flammable liquid and vapor.
Causes serious eye irritation.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 216-374-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only