missing translation for 'onlineSavingsMsg'
Learn More

1-Ethoxy-2-propanol, 90+%, remainder 2-ethoxy-1-propanol

Catalog No. p-7058817
Change view
Click to view available options
Quantity:
500 mL
4 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43306AP 500 mL
AA43306AY 4 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43306AP Supplier Thermo Scientific Chemicals Supplier No. 043306AP
Only null left

CAS: 1569-02-4 | C5H12O2 | 104.15 g/mol

1-Ethoxy-2-propanol is used as a solvent for a wide variety of resins such as epoxies, acrylic, alkyds, polyesters, nitrocellulose, and polyurethanes. It is also used in the surface coating and printing industries, thereby it regulate the flow, levelling, and coalescence of both surface coatings. It acts as an intermediate in the production of agro-chemicals and de-icing, and anti-icing formulations. It finds application as to make cleaning products, as grease and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1569-02-4
Molecular Formula C5H12O2
Molecular Weight (g/mol) 104.15
MDL Number MFCD00067050
InChI Key JOLQKTGDSGKSKJ-UHFFFAOYNA-N
Synonym 1-ethoxy-2-propanol, propylene glycol monoethyl ether, 2-propanol, 1-ethoxy, propylene glycol ethyl ether, 1-ethoxy-propan-2-ol, arcosolv r pe, 1-ethoxy-2-propan, 1-ethoxy-2-propano, acmc-1byqk, 2-hydroxypropylethylether
PubChem CID 15287
IUPAC Name 1-ethoxypropan-2-ol
SMILES CCOCC(C)O

Specifications

Melting Point -100°C
Color Colorless
Density 0.896 g/mL
Boiling Point 132°C
Flash Point 42°C (108°F)
Refractive Index 1.4065
Linear Formula CH3CH2OCH2CH(OH)CH3
Quantity 500 mL
UN Number UN1987
Solubility Information Miscible with water.
Formula Weight 104.15
Percent Purity ≥90%
Physical Form Liquid
Assay Remainder 2-ethoxy-1-propanol
Chemical Name or Material 1-Ethoxy-2-propanol
Show More Show Less
Hazard Category H226-H336
Hazard Statement GHS H Statement
H226-H336
Flammable liquid and vapor.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 216-374-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only