missing translation for 'onlineSavingsMsg'
Learn More

1-Eth-1-ynyl-4-hexylbenzene, 97%, Thermo Scientific™

Catalog No. BTB09902EA
Change view
Click to view available options
Quantity:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09902EA 10g
BTB09902EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB09902EA Supplier Thermo Scientific Chemicals Supplier No. BTB09902EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 79887-11-9
Molecular Formula C14H18
Molecular Weight (g/mol) 186.30
MDL Number MFCD00173883
InChI Key NFPDFDTYANKKIU-UHFFFAOYSA-N
Synonym 1-eth-1-ynyl-4-hexylbenzene, 4-hexyl-1-ethynylbenzene, 4-hexylphenylacetylene, 1-ethynyl-4-hexyl-benzene, 4-ethynyl-1-hexylbenzene, acmc-209pie, 4-n-hexylphenylacetylene, 1-ethynyl-4-n-hexylbenzene
PubChem CID 2775129
IUPAC Name 1-ethynyl-4-hexylbenzene
SMILES CCCCCCC1=CC=C(C=C1)C#C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula HCidentical toC6H4(CH2)5CH3
Quantity 10g
Formula Weight 186.3
Percent Purity 97%
Chemical Name or Material 1-Eth-1-ynyl-4-hexylbenzene
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.