missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosene 99.5+%, TCI America™

Catalog No. S03501ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S03501ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S03501ML Supplier TCI America Supplier No. S03501ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7-1-3452
Molecular Formula C20H40
Molecular Weight (g/mol) 280.54
MDL Number MFCD00009013
InChI Key VAMFXQBUQXONLZ-UHFFFAOYSA-N
Synonym 1-eicosene, 1-icosene, eicosene, cetyl ethylene, .alpha.-eicosene, unii-6bhh7zd637, arachidene, arachydene, cosen, alpha-eicosene
PubChem CID 18936
IUPAC Name icos-1-ene
SMILES CCCCCCCCCCCCCCCCCCC=C

Specifications

Color White
Boiling Point 151°C
Quantity 1 mL
Formula Weight 280.54
Percent Purity ≥99.5% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Eicosene [Standard Material for GC]
EINECSNumber (2)-0027
TSCA Yes