missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol 95.0+%, TCI America™

$78.29 - $527.52

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E000425G
View Documents
TCI America
E000425G
25 g N/A
Only null left
 
E0004500G
View Documents
TCI America
E0004500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

629-96-9
298.56
BTFJIXJJCSYFAL-UHFFFAOYSA-N
12404
icosan-1-ol
C20H42O
MFCD00002938
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
CHEBI:75627
CCCCCCCCCCCCCCCCCCCCO

Specifications

629-96-9
White
C20H42O
25 g
BTFJIXJJCSYFAL-UHFFFAOYSA-N
icosan-1-ol
12404
298.56
Crystalline Powder
65°C
190°C
MFCD00002938
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
CCCCCCCCCCCCCCCCCCCCO
298.56
CHEBI:75627
≥95.0% (GC)
1-Eicosanol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'