missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol 95.0+%, TCI America™

Catalog No. E000425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E000425G 25 g
E0004500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E000425G Supplier TCI America Supplier No. E000425G
Only null left

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 65°C
Color White
Boiling Point 190°C
Quantity 25 g
Formula Weight 298.56
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Eicosanol
EINECSNumber (2)-0217
TSCA Yes