missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol, 98%

CAS: 629-96-9 | C20H42O | 298.56 g/mol

$183.34 - $183.34

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC215460250
SDS View Documents
Thermo Scientific Chemicals
215460250
25 g Glass bottle
Each for $183.34
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

629-96-9
298.56
BTFJIXJJCSYFAL-UHFFFAOYSA-N
12404
icosan-1-ol
C20H42O
MFCD00002938
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
CHEBI:75627
CCCCCCCCCCCCCCCCCCCCO

Specifications

629-96-9
100.0
White
Authentic
Glass bottle
CH3(CH2)19OH
25 g
1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
BTFJIXJJCSYFAL-UHFFFAOYSA-N
icosan-1-ol
12404
298.54
Pellets, Powder or Chunks
97.5
64°C to 66°C
194°C
97.5% min. (GC)
C20H42O
MFCD00002938
01, 431
Solubility in water: slightly soluble
CCCCCCCCCCCCCCCCCCCCO
298.56
CHEBI:75627
98%
1-Eicosanol
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-119-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only