Learn More
1-Eicosanol, 96%
CAS: 629-96-9 | C20H42O | 298.56 g/mol
$71.14 - $201.25
Chemical Identifiers
| CAS | 629-96-9 |
|---|---|
| Molecular Formula | C20H42O |
| Molecular Weight (g/mol) | 298.56 |
| MDL Number | MFCD00002938 |
| InChI Key | BTFJIXJJCSYFAL-UHFFFAOYSA-N |
| Synonym | 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol |
| PubChem CID | 12404 |
| ChEBI | CHEBI:75627 |
| IUPAC Name | icosan-1-ol |
| SMILES | CCCCCCCCCCCCCCCCCCCCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0880714
|
Thermo Scientific Chemicals
L0880714 |
25 g |
Each for $71.14
|
|
|||||
|
AAL0880722
|
Thermo Scientific Chemicals
L0880722 |
100 g |
Each for $201.25
|
|
|||||
Description
1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol?s high molecular weight alcohols (policosanol?s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol′s high molecular weight alcohols (policosanol′s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.
Solubility
Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 629-96-9 | |
| 298.56 | |
| BTFJIXJJCSYFAL-UHFFFAOYSA-N | |
| 12404 | |
| icosan-1-ol |
| C20H42O | |
| MFCD00002938 | |
| 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol | |
| CHEBI:75627 | |
| CCCCCCCCCCCCCCCCCCCCO |
Specifications
| 629-96-9 | |
| 0.841 | |
| 195°C (383°F) | |
| MFCD00002938 | |
| 1705104 | |
| Insoluble in water. | |
| CCCCCCCCCCCCCCCCCCCCO | |
| 298.56 | |
| CHEBI:75627 | |
| 1-Eicosanol |
| 62°C to 66°C | |
| 220°C to 225°C (3 mmHg) | |
| C20H42O | |
| 25 g | |
| 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol | |
| BTFJIXJJCSYFAL-UHFFFAOYSA-N | |
| icosan-1-ol | |
| 12404 | |
| 96% |
Safety and Handling
EINECSNumber : 211-119-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only