missing translation for 'onlineSavingsMsg'
Learn More

1-Eicosanol, 96%

Catalog No. AAL0880714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0880714 25 g
AAL0880722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0880714 Supplier Thermo Scientific Chemicals Supplier No. L0880714
Only null left

CAS: 629-96-9 | C20H42O | 298.56 g/mol

1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol?s high molecular weight alcohols (policosanol?s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanol has been used in gradient HPLC-charged aerosol detection method for the detection of different lipids, as internal standard during determination of policosanol′s high molecular weight alcohols (policosanol′s fatty alcohols) in fish oil by GC. Also used as a non-ionic surfactant in cosmetics, used in lubricants, rubber, plastics, textiles and as an emulsion stabilizer and viscosity increasing agent.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 629-96-9
Molecular Formula C20H42O
Molecular Weight (g/mol) 298.56
MDL Number MFCD00002938
InChI Key BTFJIXJJCSYFAL-UHFFFAOYSA-N
Synonym 1-eicosanol, arachidyl alcohol, n-eicosanol, arachidic alcohol, arachic alcohol, eicosyl alcohol, eicosan-1-ol, eicosanol, 1-icosanol, n-1-eicosanol
PubChem CID 12404
ChEBI CHEBI:75627
IUPAC Name icosan-1-ol
SMILES CCCCCCCCCCCCCCCCCCCCO

Specifications

Melting Point 62°C to 66°C
Density 0.841
Boiling Point 220°C to 225°C (3 mmHg)
Flash Point 195°C (383°F)
Quantity 25 g
Beilstein 1705104
Solubility Information Insoluble in water.
Percent Purity 96%
Chemical Name or Material 1-Eicosanol
EINECSNumber 211-119-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only