missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylpyridinium Chloride 98.0+%, TCI America™

Catalog No. p-7126543
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D099525G 25 g
D0995100G 100 g
D0995500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D099525G Supplier TCI America Supplier No. D099525G
Only null left

Chemical Identifiers

CAS 104-74-5
Molecular Formula C17H30ClN
Molecular Weight (g/mol) 283.884
MDL Number MFCD00011987
InChI Key GKQHIYSTBXDYNQ-UHFFFAOYSA-M
Synonym 1-Laurylpyridinium Chloride
PubChem CID 7717
IUPAC Name 1-dodecylpyridin-1-ium;chloride
SMILES CCCCCCCCCCCC[N+]1=CC=CC=C1.[Cl-]

Specifications

Quantity 25 g
Formula Weight 283.88
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecylpyridinium Chloride
RTECSNumber UU4017070
TSCA Yes