missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Dodecylpyridinium Chloride 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Formula Weight | 283.88 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1-Dodecylpyridinium Chloride |
Safety and Handling
| RTECSNumber | UU4017070 |
| TSCA | Yes |