missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylhydantoin 98.0+%, TCI America™

Catalog No. D205225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D205225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D205225G Supplier TCI America Supplier No. D205225G
Only null left

Chemical Identifiers

CAS 85391-28-2
Molecular Formula C15H28N2O2
Molecular Weight (g/mol) 268.401
MDL Number MFCD00142675
InChI Key IHWNCNSXOZBQBX-UHFFFAOYSA-N
Synonym 1-Laurylhydantoin
PubChem CID 3020628
IUPAC Name 1-dodecylimidazolidine-2,4-dione
SMILES CCCCCCCCCCCCN1CC(=O)NC1=O

Specifications

Melting Point 100°C
Color White
Quantity 25 g
Formula Weight 268.40
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecylhydantoin