missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecylamine hydrochloride, 97%

Catalog No. AAL1231409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1231409 10 g
AAL1231418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1231409 Supplier Thermo Scientific Chemicals Supplier No. L1231409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 929-73-7
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00042017
InChI Key TWFQJFPTTMIETC-UHFFFAOYSA-N
Synonym dodecylamine hydrochloride, laurylamine hydrochloride, n-dodecylamine hydrochloride, dodecan-1-amine hydrochloride, 1-dodecanamine, hydrochloride, dodecylamine, hydrochloride, unii-1v1470j2mf, 1-dodecanamine, hydrochloride 1:1, armeen 12d hydrochloride, acmc-209rjd
PubChem CID 458426
IUPAC Name dodecan-1-amine;hydrochloride
SMILES [Cl-].CCCCCCCCCCCC[NH3+]

Specifications

Melting Point 186°C to 188°C
Linear Formula CH3(CH2)11NH2·HCl
Quantity 10 g
Beilstein 3553943
Formula Weight 221.82
Percent Purity 97%
Chemical Name or Material 1-Dodecylamine hydrochloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 213-205-7
RTECSNumber JR6750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only