missing translation for 'onlineSavingsMsg'
Learn More

(1-Dodecyl)trimethylammonium chloride, 97%

Catalog No. p-7023907
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1792414 25 g
AAA1792422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1792414 Supplier Thermo Scientific Chemicals Supplier No. A1792414
Only null left

CAS: 112-00-5 | C15H34ClN | 263.894 g/mol

Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dodecyltrimethylammonium Chloride is useful as a paint stripper, a foaming stabilizer, and a bactericidal lotion. It is used in coating, plastics, paints, rubber and ink production, as anti-static agent, catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.

Chemical Identifiers

CAS 112-00-5
Molecular Formula C15H34ClN
Molecular Weight (g/mol) 263.894
MDL Number MFCD00041974
InChI Key DDXLVDQZPFLQMZ-UHFFFAOYSA-M
Synonym dodecyltrimethylammonium chloride, alicop, cation bb, aliquat 4, n,n,n-trimethyldodecan-1-aminium chloride, laurtrimonium chloride, dehyquart lt, cation fb, nissan cation bb, lauryltrimethylammonium chloride
PubChem CID 8152
IUPAC Name dodecyl(trimethyl)azanium;chloride
SMILES CCCCCCCCCCCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point ∼237°C (decomposition)
Odor Odorless
Linear Formula CH3(CH2)11N(CH3)3Cl
Quantity 25 g
UN Number UN3077
Beilstein 3915951
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 263.9
Percent Purity 97%
Chemical Name or Material (1-Dodecyl)trimethylammonium chloride
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 203-927-0
RTECSNumber JR2122500
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only