missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecyl isocyanate, 98%

Catalog No. p-7046180
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1338003 1 g
AAL1338006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1338003 Supplier Thermo Scientific Chemicals Supplier No. L1338003
Only null left

CAS: 4202-38-4 | C13H25NO | 211.349 g/mol

1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecyl isocyanate, is incorporated in AZT analogue preparation through it use ine the synthesis of carbodiimides from phosphinimines.

Solubility
It hydrolyzes in water.

Notes
Moisture Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 4202-38-4
Molecular Formula C13H25NO
Molecular Weight (g/mol) 211.349
MDL Number MFCD00041744
InChI Key YIDSTEJLDQMWBR-UHFFFAOYSA-N
Synonym dodecyl isocyanate, dodecane, 1-isocyanato, dodecylisocyanate, 1-dodecyl isocyanate, n-dodecyl isocyanate, acmc-209jmx, isocyanic acid dodecyl ester
PubChem CID 77873
IUPAC Name 1-isocyanatododecane
SMILES CCCCCCCCCCCCN=C=O

Specifications

Density 0.877
Boiling Point 157°C to 161°C (21 mmHg)
Flash Point 134°C (273°F)
Odor Irritating
Refractive Index 1.441
Quantity 1 g
UN Number UN2206
Beilstein 1774823
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 211.35
Percent Purity 98%
Chemical Name or Material 1-Dodecyl isocyanate
Show More Show Less
Hazard Category H302+H332-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P332+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 224-111-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only