missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecene 95.0+%, TCI America™

Catalog No. p-7126533
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D097425ML 25 mL
D0974500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D097425ML Supplier TCI America Supplier No. D097425ML
Only null left

Chemical Identifiers

CAS 112-41-4
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00008961
InChI Key CRSBERNSMYQZNG-UHFFFAOYSA-N
Synonym 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
PubChem CID 8183
IUPAC Name dodec-1-ene
SMILES CCCCCCCCCCC=C

Specifications

Melting Point -35°C
Color Colorless
Boiling Point 213°C
Quantity 25 mL
UN Number 3082
Formula Weight 168.32
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Dodecene
EINECSNumber (2)-0027
RTECSNumber JR5120000
TSCA Yes