missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecene, 96%

Catalog No. p-7022287
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1499222 100 g
AAA1499236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1499222 Supplier Thermo Scientific Chemicals Supplier No. A1499222

CAS: 112-41-4 | C12H24 | 168.324 g/mol

1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Dodecene, is used in Oil and Gas field drilling and production operations, in polymer production, as an intermediate, coating, cleaning agent, lubricants and also used as a functional fluid. It is also used in the production of detergents.

Solubility
It is soluble in ethanol, ethyl ether, and acetone

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from bases, oxidizing agents, reducing agents.

Chemical Identifiers

CAS 112-41-4
Molecular Formula C12H24
Molecular Weight (g/mol) 168.324
MDL Number MFCD00008961
InChI Key CRSBERNSMYQZNG-UHFFFAOYSA-N
Synonym 1-dodecene, dodecene, adacene 12, n-dodec-1-ene, dodecylene, alpha-dodecene, alpha-dodecylene, dodecene-1, .alpha.-dodecene, unii-wye669f3gr
PubChem CID 8183
IUPAC Name dodec-1-ene
SMILES CCCCCCCCCCC=C

Specifications

Melting Point -37°C
Density 0.758
Boiling Point 212°C to 213°C
Flash Point 73°C (163°F)
Refractive Index 1.43
Linear Formula CH3(CH2)9CH=CH2
Quantity 100 g
Beilstein 1699848
Solubility Information It is soluble in ethanol,ethyl ether,and acetone
Formula Weight 168.32
Percent Purity 96%
Chemical Name or Material 1-Dodecene
Show More Show Less
Hazard Category H227-H304
Hazard Statement GHS H Statement
H304-H315-H227
May be fatal if swallowed and enters airways.
Causes skin irritation.
Combustible liquid.
Precautionary Statement P210-P280-P301+P310-P331-P370+P378q-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 203-968-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only