missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanethiol 95.0+%, TCI America™

Catalog No. D0970500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0970500ML 500 mL
D097025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0970500ML Supplier TCI America Supplier No. D0970500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00004885
InChI Key WNAHIZMDSQCWRP-UHFFFAOYSA-N
Synonym 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
PubChem CID 8195
IUPAC Name dodecane-1-thiol
SMILES CCCCCCCCCCCCS

Specifications

Melting Point -7°C
Color Yellow
Boiling Point 277°C
Quantity 500 mL
UN Number 1760
Formula Weight 202.40
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Dodecanethiol
EINECSNumber (2)-0464
RTECSNumber JR3155000
TSCA Yes