missing translation for 'onlineSavingsMsg'
Learn More

1-Docosene 99.0+%, TCI America™

Catalog No. D101325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D101325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D101325G Supplier TCI America Supplier No. D101325G
Only null left

Chemical Identifiers

CAS 1599-67-3
Molecular Formula C22H44
Molecular Weight (g/mol) 308.59
MDL Number MFCD00027085
InChI Key SPURMHFLEKVAAS-UHFFFAOYSA-N
PubChem CID 74138
ChEBI CHEBI:84220
IUPAC Name docos-1-ene
SMILES CCCCCCCCCCCCCCCCCCCCC=C

Specifications

Color White
Quantity 25 g
Formula Weight 308.59
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material 1-Docosene
EINECSNumber (2)-0027
TSCA Yes