missing translation for 'onlineSavingsMsg'
Learn More

1-Dimethylamino-2-nitroethylene, 98%

Catalog No. AAA1163703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1163703 1 g
AAA1163706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1163703 Supplier Thermo Scientific Chemicals Supplier No. A1163703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1190-92-7 | C4H8N2O2 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1190-92-7
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00051519
InChI Key JKOVQYWMFZTKMX-ONEGZZNKSA-N
Synonym 1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine
PubChem CID 637928
IUPAC Name (E)-N,N-dimethyl-2-nitroethenamine
SMILES CN(C)C=C[N+](=O)[O-]

Specifications

Melting Point 101°C to 105°C
Quantity 1 g
Beilstein 1903136
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material 1-Dimethylamino-2-nitroethylene
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only