missing translation for 'onlineSavingsMsg'
Learn More

(+)-1-Deoxynojirimycin

CAS: 19130-96-2 | C6H13NO4 | 163.17 g/mol

Supplier:  Thermo Scientific Chemicals J62602MC

Catalog No. AAJ62602MC


Only null left

Description

Description

An alpha-glucosidase inhibitor. Interferes with normal processing of N-linked glycoproteins.(+)-1-Deoxynojirimycin acts as an inhibitor of alfa-glucosidase I and II and maltase-glucoamylase. It also inhibits mammalian glucosidase, intestinal and lysosmal, beta-glucosidase from sweet almonds, pancreatic alfa-amylase and amyloglucosidase. Further, it serves as a enzyme enhancer for the treatment of Fabry and Pompe disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong acids and strong bases.
Specifications

Specifications

(+)-1-Deoxynojirimycin
Powder
195°C to 196°C
100 mg
MFCD00063474
1,5-Dideoxy-1,5-imino-D-glucitol; (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
LXBIFEVIBLOUGU-JGWLITMVSA-N
(2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
29435
163.17
(+)-1-Deoxynojirimycin
19130-96-2
White
C6H13NO4
142905
Soluble in water,dimethyl sulfoxide and methanol.
OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
163.17
CHEBI:44369

Chemical Identifiers

19130-96-2
163.17
LXBIFEVIBLOUGU-JGWLITMVSA-N
29435
(2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
C6H13NO4
MFCD00063474
1,5-Dideoxy-1,5-imino-D-glucitol; (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
CHEBI:44369
OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : TN4350300

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only