missing translation for 'onlineSavingsMsg'
Learn More

1-Deoxynojirimycin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013413
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013413 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013413 Supplier Merck Emd Millipore Supplier No. 0801210MG
Only null left

1-Deoxynojirimycin (DNJ) is a type of polyhydroxylated piperidine alkaloid that is a glucosidase inhibitor.

Specifications

CAS 19130-96-2
Linear Formula C6H13 NO4
MDL Number MFCD00063474
Synonym 1,5-Dideoxy-1,5-imino-D-sorbitol
Type Analytical Standard
Quantity 10 mg
Formula Weight 163.17
Percent Purity ≥95% (HPLC)
Grade Analytical Standard