missing translation for 'onlineSavingsMsg'
Learn More

1-Decyne, 98%

CAS: 764-93-2 | C10H18 | 138.25 g/mol

$611.24 - $611.24

Chemical Identifiers

CAS 764-93-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00009576
InChI Key ILLHQJIJCRNRCJ-UHFFFAOYSA-N
Synonym 1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
PubChem CID 12997
ChEBI CHEBI:87322
IUPAC Name dec-1-yne
SMILES CCCCCCCCC#C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC223431000
SDS View Documents
Thermo Scientific Chemicals
223431000
100 mL Glass bottle
Each for $611.24
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

764-93-2
138.25
ILLHQJIJCRNRCJ-UHFFFAOYSA-N
12997
dec-1-yne
C10H18
MFCD00009576
1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
CHEBI:87322
CCCCCCCCC#C

Specifications

764-93-2
100.0
Colorless to Yellow
174.0°C
Authentic
Glass bottle
1.4260 to 1.4280
MFCD00009576
01,I,122
1-decyne, decyne, octylacetylene, unii-ulr28gd98q, ulr28gd98q, decine, n-octylacetylene, deca-1-yne, acmc-209p3y
CCCCCCCCC#C
138.25
CHEBI:87322
98%
1-Decyne, 98%
97.5
-44.0°C
0.7600g/mL
48°C
97.5% min. (GC)
C10H18
CH3(CH2)7C≡CH
100 mL
0.76
ILLHQJIJCRNRCJ-UHFFFAOYSA-N
dec-1-yne
12997
138.25
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 212-132-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only