missing translation for 'onlineSavingsMsg'
Learn More

1-Decylboronic acid, 98%

CAS: 24464-63-9 | C10H23BO2 | 186.102 g/mol

Supplier:  Thermo Scientific Chemicals L1995714

 View more versions of this product

Catalog No. AAL1995714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1-Decylboronic acid
86°C to 89°C
MFCD01074638
1743934
TUTSEMMBKGBUGD-UHFFFAOYSA-N
decylboronic acid
4186635
98%
24464-63-9
C10H23BO2
25 g
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d
B(CCCCCCCCCC)(O)O
186.102
186.1

Chemical Identifiers

24464-63-9
186.102
TUTSEMMBKGBUGD-UHFFFAOYSA-N
4186635
B(CCCCCCCCCC)(O)O
C10H23BO2
MFCD01074638
n-decylboronic acid, boronic acid, decyl, decylboranic acid, n-decaneboronic acid, n-, boronicacid, b-decyl, 1-decylboronic acid, ksc199m8d
decylboronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only