missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
$253.51 - $831.37
Chemical Identifiers
| CAS | 433337-23-6 |
|---|---|
| Molecular Formula | C16H27F6N3O4S2 |
| Molecular Weight (g/mol) | 503.519 |
| MDL Number | MFCD12022376 |
| InChI Key | ZYVGZWFCGPUVSH-UHFFFAOYSA-N |
| PubChem CID | 12040509 |
| IUPAC Name | bis(trifluoromethylsulfonyl)azanide;1-decyl-3-methylimidazol-3-ium |
| SMILES | CCCCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 433337-23-6 | |
| 503.519 | |
| ZYVGZWFCGPUVSH-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-decyl-3-methylimidazol-3-ium |
| C16H27F6N3O4S2 | |
| MFCD12022376 | |
| 12040509 | |
| CCCCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F |
Specifications
| 433337-23-6 | |
| Yellow | |
| MFCD12022376 | |
| ZYVGZWFCGPUVSH-UHFFFAOYSA-N | |
| bis(trifluoromethylsulfonyl)azanide;1-decyl-3-methylimidazol-3-ium | |
| 12040509 | |
| ≥98.0% (HPLC,T) | |
| 1-Decyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide |
| -19°C | |
| C16H27F6N3O4S2 | |
| 5 g | |
| CCCCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F | |
| 503.519 | |
| 503.52 | |
| Liquid |
Safety and Handling
TSCA : No