missing translation for 'onlineSavingsMsg'
Learn More

1-Decanethiol 95.0+%, TCI America™

Catalog No. p-7125984
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D001625ML 25 mL
D0016100ML 100 mL
D0016500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D001625ML Supplier TCI America Supplier No. D001625ML
Only null left

Chemical Identifiers

CAS 143-10-2
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.346
MDL Number MFCD00004884
InChI Key VTXVGVNLYGSIAR-UHFFFAOYSA-N
Synonym 1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol
PubChem CID 8917
IUPAC Name decane-1-thiol
SMILES CCCCCCCCCCS

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 241°C
Quantity 25 mL
Formula Weight 174.35
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Decanethiol
EINECSNumber (2)-0464
TSCA Yes