missing translation for 'onlineSavingsMsg'
Learn More

1-Decanethiol 95.0+%, TCI America™

Catalog No. d001625ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
D001625ML 25 mL
D0016100ML 100 mL
D0016500ML 500 mL
3 options

Catalog No. D001625ML

Supplier: TCI America D001625ML

Only null left

Chemical Identifiers

CAS 143-10-2
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.346
MDL Number MFCD00004884
InChI Key VTXVGVNLYGSIAR-UHFFFAOYSA-N
Synonym 1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol
PubChem CID 8917
IUPAC Name decane-1-thiol
SMILES CCCCCCCCCCS

Specifications

Melting Point -26°C
Color Yellow
Boiling Point 241°C
Quantity 25 mL
Formula Weight 174.35
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Decanethiol
EINECSNumber (2)-0464
TSCA Yes