missing translation for 'onlineSavingsMsg'
Learn More

1-Decanethiol, 96%

Catalog No. AAA15913AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15913AC 25 mL
AAA15913AE 100 mL
AAA15913AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA15913AC Supplier Thermo Scientific Chemicals Supplier No. A15913AC
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 143-10-2 | C10H22S | 174.346 g/mol

1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanethiol is used to improve charge injection by creating a monolayer of this material on gold source-drain electrode surfaces for field-effect transistors (FETs). It is also used in the study of self-assembled monolayers.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. It is sensitive to air. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 143-10-2
Molecular Formula C10H22S
Molecular Weight (g/mol) 174.346
MDL Number MFCD00004884
InChI Key VTXVGVNLYGSIAR-UHFFFAOYSA-N
Synonym 1-decanethiol, decyl mercaptan, 1-mercaptodecane, decylmercaptan, decanethiol, decanethiol-1, decanthiol, n-decanethiol, n-decanthiol, 1-decylthiol
PubChem CID 8917
IUPAC Name decane-1-thiol
SMILES CCCCCCCCCCS

Specifications

Melting Point -26°C
Density 0.843
Boiling Point 237°C
Flash Point 98°C (208°F)
Odor Unpleasant
Refractive Index 1.4565
Linear Formula CH3(CH2)9SH
Quantity 25 mL
Beilstein 1735223
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 174.35
Percent Purity 96%
Chemical Name or Material 1-Decanethiol
Show More Show Less
Hazard Category H318-H335
Hazard Statement GHS H Statement
H318-H335
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 205-584-2
TSCA Yes
Recommended Storage Store under Argon

RUO – Research Use Only