missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic Acid Sodium Salt, Approx. 99%, Spectrum™ Chemical

Catalog No. 18608097
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608097 5 g Amber Glass Bottle
18608098 25 g Amber Glass Bottle
18608099 100 g Amber Glass Bottle
18608100 1 kg Poly Pail
18608101 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18608097 Supplier Spectrum Chemical Mfg Cor Supplier No. D20045GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 13419-61-9

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity ∼99%
Grade Reagent

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.