missing translation for 'onlineSavingsMsg'
Learn More

1-Decanesulfonic acid, sodium salt, 96%

Catalog No. p-2689532
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC369450250 25 g Glass bottle
AC369451000 100 g Glass bottle
2 options

Catalog No. AC369450250

Supplier: Thermo Scientific Chemicals 369450250

Only null left

1-Decanesulfonic acid, sodium salt, 96%, Quantity: 25g, Packaging: Glass bottle, White to Cream, Melting Point: >300 deg.C, Molecular Weight: 244.32, Percent Purity: 96%, Assay Percent Range: 96%, Beilstein: 04, III, 27, CAS: 13419-61-9, CAS Max %: 100.0, CAS Min %: 95.0 | CAS: 13419-61-9 | C10H21NaO3S | 244.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13419-61-9
Molecular Formula C10H21NaO3S
Molecular Weight (g/mol) 244.32
MDL Number MFCD00007526
InChI Key AIMUHNZKNFEZSN-UHFFFAOYSA-M
Synonym sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt
PubChem CID 2724181
IUPAC Name sodium;decane-1-sulfonate
SMILES CCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point >300°C
Color White to Cream
Loss on Drying 5% max. (100°C, 3 hrs) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3(CH2)9SO3Na
Quantity 25 g
Beilstein 04, III, 27
Solubility Information Solubility in water: soluble
Formula Weight 244.32
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 1-Decanesulfonic acid, sodium salt, 96%
Show More Show Less
EINECSNumber 236-525-9

RUO – Research Use Only