missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclopropylcarbonyl)piperazine Hydrochloride 98.0+%, TCI America™

Catalog No. C31125G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31125G 5 g
C31121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31125G Supplier TCI America Supplier No. C31125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1021298-67-8
Molecular Formula C8H15ClN2O
Molecular Weight (g/mol) 190.67
MDL Number MFCD03428576
InChI Key WIHDAPMHNYYTOA-UHFFFAOYSA-N
Synonym 1-cyclopropylcarbonyl piperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hydrochloride, 1-cyclopropanecarbonylpiperazine hydrochloride, cyclopropyl piperazin-1-yl methanone hcl, cyclopropylcarboxylic acid-1-piperazineamide hydrochloride, cyclopropyl piperazinyl ketone, chloride, pubchem19546, 1-cyclopropylcarbonyl piperazine hcl, 1-cyclopropylcarbonyl-piperazine hcl, 1-cyclopropylcarbonyl piperazine hcl salt
PubChem CID 20847955
IUPAC Name 1-cyclopropanecarbonylpiperazine hydrochloride
SMILES Cl.O=C(C1CC1)N1CCNCC1

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine Hydrochloride