missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid 98.0+%, TCI America™
$124.89 - $124.89
Chemical Identifiers
| CAS | 93107-30-3 |
|---|---|
| Molecular Formula | C13H9F2NO3 |
| Molecular Weight (g/mol) | 265.216 |
| MDL Number | MFCD01646375 |
| InChI Key | KNEXGVPHPGXAGF-UHFFFAOYSA-N |
| PubChem CID | 5210422 |
| IUPAC Name | 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid |
| SMILES | C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)F)F)C(=O)O |
Chemical Identifiers
| 93107-30-3 | |
| 265.216 | |
| KNEXGVPHPGXAGF-UHFFFAOYSA-N | |
| 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid |
| C13H9F2NO3 | |
| MFCD01646375 | |
| 5210422 | |
| C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)F)F)C(=O)O |
Specifications
| 93107-30-3 | |
| White | |
| MFCD01646375 | |
| KNEXGVPHPGXAGF-UHFFFAOYSA-N | |
| 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid | |
| 5210422 | |
| ≥98.0% (HPLC,T) | |
| 1-Cyclopropyl-6,7-difluoro-1,4-dihydro-4-oxoquinoline-3-carboxylic Acid |
| 289°C | |
| C13H9F2NO3 | |
| 1 g | |
| C1CC1N2C=C(C(=O)C3=CC(=C(C=C32)F)F)C(=O)O | |
| 265.216 | |
| 265.22 | |
| Crystalline Powder |
Safety and Handling
TSCA : No