missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentylethanol, 97%

Catalog No. AAL0344604
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0344604 2 g
AAL0344609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0344604 Supplier Thermo Scientific Chemicals Supplier No. L0344604

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 52829-98-8 | C7H14O | 114.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52829-98-8
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00013743
InChI Key WPCMSUSLCWXTKB-UHFFFAOYNA-N
Synonym 1-cyclopentylethan-1-ol, cyclopentanemethanol, .alpha.-methyl, alpha-methylcyclopentanemethanol, cyclopentyl ethanol, 1-hydroxyethylcyclopentane, 1-cyclopentylethanol, cyclopentanemethanol, alpha-methyl
PubChem CID 95434
IUPAC Name 1-cyclopentylethanol
SMILES CC(O)C1CCCC1

Specifications

Density 0.911
Boiling Point 68°C (8 mmHg)
Refractive Index 1.458
Quantity 2 g
Beilstein 2069851
Formula Weight 114.19
Percent Purity 97%
Chemical Name or Material 1-Cyclopentylethanol
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 258-208-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only