missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenecarboxylic Acid 98.0+%, TCI America™

$158.44 - $630.05

Chemical Identifiers

CAS 1560-11-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00001396
InChI Key PYRZPBDTPRQYKG-UHFFFAOYSA-N
Synonym 1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
PubChem CID 95964
IUPAC Name cyclopentene-1-carboxylic acid
SMILES C1CC=C(C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C17021G
View Documents
TCI America
C17021G
1 g N/A
Only null left
 
C17025G
View Documents
TCI America
C17025G
5 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

1560-11-8
112.128
PYRZPBDTPRQYKG-UHFFFAOYSA-N
95964
C1CC=C(C1)C(=O)O
C6H8O2
MFCD00001396
1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
cyclopentene-1-carboxylic acid

Specifications

1560-11-8
White-Yellow
MFCD00001396
3261
PYRZPBDTPRQYKG-UHFFFAOYSA-N
cyclopentene-1-carboxylic acid
95964
≥98.0% (HPLC,T)
1-Cyclopentenecarboxylic Acid
122°C
C6H8O2
1 g
1-cyclopentenecarboxylic acid, isoaleprolic acid, 1-cyclopentene-1-carboxylic acid, cyclopent-1-ene-1-carboxylic acid, cyclopent-1-enecarboxylic acid, cyclopentenoic acid, 1-cyclopentenylcarboxylic acid, cyclopentenecarboxylic acid, acmc-1bxxw, cyclopentaencarboxylic acid
C1CC=C(C1)C(=O)O
112.128
112.13
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : GY5976100

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'