missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclohexylmethyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAH5549103
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5549103 1 g
AAH55491MD 250 mg
AAH5549106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5549103 Supplier Thermo Scientific Chemicals Supplier No. H5549103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 57184-23-3
Molecular Formula C11H22N2
Molecular Weight (g/mol) 182.31
MDL Number MFCD01075237
InChI Key LRPGNFROBDUREU-UHFFFAOYSA-N
PubChem CID 2735885
IUPAC Name 1-(cyclohexylmethyl)piperazine
SMILES C(C1CCCCC1)N1CCNCC1

Specifications

Color Colorless
Density 0.938 g/mL
Boiling Point 230°C to 231°C
Flash Point 87°C (189°F)
Refractive Index 1.495
Quantity 1 g
Synonym 1-cyclohexylmethyl piperazine, piperazine, 1-cyclohexylmethyl, cyclohexylmethyl piperazine, 1-cyclohexylmethyl-piperazine, 1-cyclohexyl-methyl piperazine, piperazine,1-cyclohexylmethyl
Formula Weight 182.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(Cyclohexylmethyl)piperazine
Show More Show Less
Hazard Category H227-H315-H318-H335
Hazard Statement GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Ambient temperatures