Learn More
1-Cyclohexylethanol, 98%
CAS: 1193-81-3 | C8H16O | 128.215 g/mol
$145.11 - $503.11
Chemical Identifiers
| CAS | 1193-81-3 |
|---|---|
| Molecular Formula | C8H16O |
| Molecular Weight (g/mol) | 128.215 |
| MDL Number | MFCD00001475 |
| InChI Key | JMSUNAQVHOHLMX-UHFFFAOYSA-N |
| Synonym | methylcyclohexylcarbinol, 1-cyclohexyl-1-ethanol, cyclohexylmethylcarbinol, 1-cyclohexylethan-1-ol, ethanol, 1-cyclohexyl, methanol, cyclohexylmethyl, cyclohexanemethanol, .alpha.-methyl, +--1-cyclohexylethanol, 1-cyclohexyl-ethanol, acmc-20aovg |
| PubChem CID | 137829 |
| IUPAC Name | 1-cyclohexylethanol |
| SMILES | CC(C1CCCCC1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2036306
|
Thermo Scientific Chemicals
B2036306 |
5 g |
Each for $145.11
|
|
|||||
|
AAB2036314
|
Thermo Scientific Chemicals
B2036314 |
25 g |
Each for $503.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1193-81-3 | |
| 128.215 | |
| JMSUNAQVHOHLMX-UHFFFAOYSA-N | |
| 137829 | |
| CC(C1CCCCC1)O |
| C8H16O | |
| MFCD00001475 | |
| methylcyclohexylcarbinol, 1-cyclohexyl-1-ethanol, cyclohexylmethylcarbinol, 1-cyclohexylethan-1-ol, ethanol, 1-cyclohexyl, methanol, cyclohexylmethyl, cyclohexanemethanol, .alpha.-methyl, +--1-cyclohexylethanol, 1-cyclohexyl-ethanol, acmc-20aovg | |
| 1-cyclohexylethanol |
Specifications
| 1193-81-3 | |
| 188°C to 190°C | |
| C8H16O | |
| MFCD00001475 | |
| methylcyclohexylcarbinol, 1-cyclohexyl-1-ethanol, cyclohexylmethylcarbinol, 1-cyclohexylethan-1-ol, ethanol, 1-cyclohexyl, methanol, cyclohexylmethyl, cyclohexanemethanol, .alpha.-methyl, +--1-cyclohexylethanol, 1-cyclohexyl-ethanol, acmc-20aovg | |
| CC(C1CCCCC1)O | |
| 128.215 | |
| 128.22 | |
| 1-Cyclohexylethanol |
| 0.92 | |
| 73°C (163°F) | |
| 1.465 | |
| 5 g | |
| JMSUNAQVHOHLMX-UHFFFAOYSA-N | |
| 1-cyclohexylethanol | |
| 137829 | |
| 98% |
Safety and Handling
GHS H Statement
H301-H311-H227
Toxic if swallowed.
Toxic in contact with skin.
Combustible liquid.
P210-P264b-P280-P302+P352-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H312-H315-H319
EINECSNumber : 214-780-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only