missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclohexylcarbonyl)piperazine 98.0+%, TCI America™

Catalog No. C33771G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33771G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C33771G Supplier TCI America Supplier No. C33771G

Chemical Identifiers

CAS 27561-62-2
Molecular Formula C11H20N2O
Molecular Weight (g/mol) 196.294
MDL Number MFCD06798113
InChI Key ZSZROXCAFYZNHE-UHFFFAOYSA-N
Synonym 1-cyclohexylcarbonyl piperazine, cyclohexyl piperazin-1-yl methanone, 1-cyclohexanecarbonylpiperazine, methanone, cyclohexyl-1-piperazinyl, maybridge1_006754, cyclohexylpiperazino ketone, cyclohexyl piperazinyl ketone, n-cyclohexylcarbonyl piperazine, cyclohexyl-1-piperazinylmethanone, 1-cyclohexanecarbonyl-piperazine
PubChem CID 3437502
IUPAC Name cyclohexyl(piperazin-1-yl)methanone
SMILES C1CCC(CC1)C(=O)N2CCNCC2

Specifications

Melting Point 88°C
Color White-Yellow
Quantity 1 g
Formula Weight 196.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Cyclohexylcarbonyl)piperazine