missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide Metho-p-toluenesulfonate 96.0+%, TCI America™

Catalog No. C07935G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C07935G 5 g
C079325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C07935G Supplier TCI America Supplier No. C07935G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2491-17-0
Molecular Formula C21H33N3O4S
Molecular Weight (g/mol) 423.57
MDL Number MFCD00011979
InChI Key GBCAVSYHPPARHX-UHFFFAOYSA-M
Synonym cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
PubChem CID 17220
IUPAC Name 4-(2-{[(cyclohexylimino)methylidene]amino}ethyl)-4-methylmorpholin-4-ium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 423.57
Percent Purity ≥96.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide Metho-p-toluenesulfonate [for Peptide Synthesis]