missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%

Catalog No. AC111360050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC111360050 5 g Glass bottle
AC111360250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC111360050 Supplier Thermo Scientific Chemicals Supplier No. 111360050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2491-17-0 | C21H33N3O4S | 423.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 2491-17-0
Molecular Formula C21H33N3O4S
Molecular Weight (g/mol) 423.57
MDL Number MFCD00011979
InChI Key GBCAVSYHPPARHX-UHFFFAOYSA-M
Synonym cme-carbodiimide, morpho cdi, 1-cyclohexyl 3-2-morpholinoethyl carbodiimide methotosylate, 4-2-cyclohexylimino methylene amino ethyl-4-methylmorpholin-4-ium 4-methylbenzenesulfonate, cmc, 1-cyclohexyl-3-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, n-cyclohexyl-n'-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, cmc n-cyclohexyl-n-2-morpholinoethyl carbodiimide metho-p-toluenesulfonate, chm van, 4-5-cyclohexyl-3,5-diazapenta-3,4-dienyl-4-methylmorpholine, 4-methylbenzene sulfonic acid
PubChem CID 17220
IUPAC Name N'-cyclohexyl-N-[2-(4-methylmorpholin-4-ium-4-yl)ethyl]methanediimine;4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.C[N+]1(CCN=C=NC2CCCCC2)CCOCC1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 110.0°C to 116.0°C
Color White to Yellow
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Fieser 01,181; 11,151
Solubility Information Solubility in water: soluble in water
Formula Weight 423.57
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Cyclohexyl-3-(2-morpholinoethyl)carbodiimide metho-p-toluenesulfonate, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 219-650-3
TSCA TSCA

RUO – Research Use Only