missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-2-pyrrolidone 99.0+%, TCI America™

Catalog No. p-7124709
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C096325ML 25 mL
C0963500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C096325ML Supplier TCI America Supplier No. C096325ML

Chemical Identifiers

CAS 6837-24-7
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00003191
InChI Key PZYDAVFRVJXFHS-UHFFFAOYSA-N
Synonym n-cyclohexyl-2-pyrrolidone, 1-cyclohexyl-2-pyrrolidone, 2-pyrrolidinone, 1-cyclohexyl, cyclohexyl pyrrolidone, n-cyclohexylpyrrolidone, 1-cyclohexyl-2-pyrrolidinone, n-cyclohexylpyrrolidinone, unii-7im8p1vvnm, 7im8p1vvnm, acmc-20aj1a
PubChem CID 81278
IUPAC Name 1-cyclohexylpyrrolidin-2-one
SMILES C1CCC(CC1)N2CCCC2=O

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 mL
UN Number 2810
Formula Weight 167.25
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Cyclohexyl-2-pyrrolidone
RTECSNumber UY5748450
TSCA Yes