missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-2-pyrrolidinone, 99%

Catalog No. AAA1663818
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1663818 50 g
AAA1663830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1663818 Supplier Thermo Scientific Chemicals Supplier No. A1663818

CAS: 6837-24-7 | C10H17NO | 167.252 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6837-24-7
Molecular Formula C10H17NO
Molecular Weight (g/mol) 167.252
MDL Number MFCD00003191
InChI Key PZYDAVFRVJXFHS-UHFFFAOYSA-N
Synonym n-cyclohexyl-2-pyrrolidone, 1-cyclohexyl-2-pyrrolidone, 2-pyrrolidinone, 1-cyclohexyl, cyclohexyl pyrrolidone, n-cyclohexylpyrrolidone, 1-cyclohexyl-2-pyrrolidinone, n-cyclohexylpyrrolidinone, unii-7im8p1vvnm, 7im8p1vvnm, acmc-20aj1a
PubChem CID 81278
IUPAC Name 1-cyclohexylpyrrolidin-2-one
SMILES C1CCC(CC1)N2CCCC2=O

Specifications

Melting Point 12°C to 15°C
Density 1.007
Boiling Point 290°C to 292°C
Flash Point 140°C (284°F)
Refractive Index 1.499
Quantity 50 g
UN Number UN2810
Beilstein 121832
Formula Weight 167.25
Percent Purity 99%
Chemical Name or Material 1-Cyclohexyl-2-pyrrolidinone
Show More Show Less
Hazard Category H302+H312-H315-H319-H330-H335
Hazard Statement GHS H Statement
H330-H318-H302-H312-H315-H335
Fatal if inhaled.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 229-919-7
RTECSNumber UY5748450
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only