missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7124901
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C12961G 1 g
C12965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C12961G Supplier TCI America Supplier No. C12961G
Only null left

Chemical Identifiers

CAS 636-82-8
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.155
MDL Number MFCD00001545
InChI Key NMEZJSDUZQOPFE-UHFFFAOYSA-N
Synonym 1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
PubChem CID 69470
ChEBI CHEBI:29565
IUPAC Name cyclohexene-1-carboxylic acid
SMILES C1CCC(=CC1)C(=O)O

Specifications

Color White-Yellow
Boiling Point 135°C
Quantity 1 g
Formula Weight 126.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Cyclohexene-1-carboxylic Acid