missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-carboxylic acid, 97%

CAS: 636-82-8 | C7H10O2 | 126.155 g/mol

Supplier:  Thermo Scientific Chemicals A1074106

 View more versions of this product

Catalog No. AAA1074106


Only null left
Explore available promotions

Description

Description

1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.

Solubility
Soluble in water (0.7 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Specifications

Specifications

1-Cyclohexene-1-carboxylic acid
30°C to 34°C
138°C to 140°C (14 mmHg)
C7H10O2
MFCD00001545
UN3261
1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
NMEZJSDUZQOPFE-UHFFFAOYSA-N
cyclohexene-1-carboxylic acid
69470
126.16
636-82-8
1.1
>110°C (230°F)
1.499
5 g
1905520
Soluble in water (0.7g/L at 20°C).
C1CCC(=CC1)C(=O)O
126.155
CHEBI:29565
97%

Chemical Identifiers

636-82-8
126.155
NMEZJSDUZQOPFE-UHFFFAOYSA-N
69470
cyclohexene-1-carboxylic acid
C7H10O2
MFCD00001545
1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid
CHEBI:29565
C1CCC(=CC1)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only