missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexene-1-carbonitrile, 98%

Catalog No. AAL1028409
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1028409 10 g
AAL1028404 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1028409 Supplier Thermo Scientific Chemicals Supplier No. L1028409

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1855-63-6 | C7H9N | 107.156 g/mol

1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Cyclohexene-1-carbonitrile has been used in the preparation of cyclohexene-1-carboxylic acid.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1855-63-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00013772
InChI Key GTMWGXABXQTZRJ-UHFFFAOYSA-N
Synonym 1-cyanocyclohexene, cyanocyclohexene, 1-cyclohexene-1-carbonitrile, cyclohexenecarbonitrile, 1-cyclohexenecarbonitrile, cyclohex-1-enecarbonitrile, cyclohex-1-ene-1-carbonitrile, 1-cyclohexenyl cyanide, 1-cyano-1-cyclohexene, acmc-1cqlm
PubChem CID 74619
IUPAC Name cyclohexene-1-carbonitrile
SMILES C1CCC(=CC1)C#N

Specifications

Density 0.947
Boiling Point 85°C to 88°C (17 mmHg)
Flash Point 83°C (181°F)
Refractive Index 1.482
Quantity 10 g
UN Number UN3276
Beilstein 385837
Solubility Information Not miscible in water.
Formula Weight 107.16
Percent Purity 98%
Chemical Name or Material 1-Cyclohexene-1-carbonitrile
Show More Show Less
Hazard Category H227-H302+H312+H332
Hazard Statement GHS H Statement
H331-H227-H302-H312
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 217-454-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only