missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyanovinyl Acetate (stabilized with TBC) 97.0+%, TCI America™
$515.03 - $1783.35
Chemical Identifiers
| CAS | 3061-65-2 |
|---|---|
| Molecular Formula | C5H5NO2 |
| Molecular Weight (g/mol) | 111.1 |
| MDL Number | MFCD00001868 |
| InChI Key | LFSHREXVLSTLFB-UHFFFAOYSA-N |
| Synonym | 2-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester |
| PubChem CID | 18291 |
| IUPAC Name | 1-cyanoethenyl acetate |
| SMILES | CC(=O)OC(=C)C#N |
Chemical Identifiers
| 3061-65-2 | |
| 111.1 | |
| LFSHREXVLSTLFB-UHFFFAOYSA-N | |
| 18291 | |
| CC(=O)OC(=C)C#N |
| C5H5NO2 | |
| MFCD00001868 | |
| 2-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester | |
| 1-cyanoethenyl acetate |
Specifications
| 3061-65-2 | |
| 173°C | |
| MFCD00001868 | |
| 3276 | |
| LFSHREXVLSTLFB-UHFFFAOYSA-N | |
| 1-cyanoethenyl acetate | |
| 18291 | |
| ≥97.0% (GC) | |
| 1-Cyanovinyl Acetate (stabilized with TBC) |
| Yellow | |
| C5H5NO2 | |
| 5 g | |
| 2-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester | |
| CC(=O)OC(=C)C#N | |
| 111.1 | |
| 111.10 | |
| Liquid |
Safety and Handling
RTECSNumber : AG4550000
TSCA : No
Recommended Storage : Refrigerator