missing translation for 'onlineSavingsMsg'
Learn More

1-Cyanomethylpiperidine 98.0+%, TCI America™

Catalog No. C131425ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C131425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C131425ML Supplier TCI America Supplier No. C131425ML

Chemical Identifiers

CAS 3010-03-5
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD00023733
InChI Key CLVBVRODHJFTGF-UHFFFAOYSA-N
Synonym piperidinoacetonitrile, piperidin-1-ylacetonitrile, n-cyanomethylpiperidine, 1-piperidineacetonitrile, piperidoacetonitrile, piperidine-1-acetonitrile, 2-piperidin-1-yl acetonitrile, 1-cyanomethylpiperidine, n-piperidinoacetonitrile, acetonitrile, piperidino
PubChem CID 18164
IUPAC Name 2-piperidin-1-ylacetonitrile
SMILES C1CCN(CC1)CC#N

Specifications

Color White-Yellow
Boiling Point 210°C
Quantity 25 mL
UN Number 3439
Formula Weight 124.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Cyanomethylpiperidine
RTECSNumber AM1925000
TSCA No