missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyanomethyl)piperidinium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. C24215G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C24215G Supplier TCI America Supplier No. C24215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 434937-12-9
Molecular Formula C7H13BF4N2
Molecular Weight (g/mol) 211.999
InChI Key HSIKBRNFKUNHIN-UHFFFAOYSA-O
PubChem CID 53384401
IUPAC Name 2-piperidin-1-ium-1-ylacetonitrile;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C1CC[NH+](CC1)CC#N

Specifications

Melting Point 105°C
Color White
Quantity 5 g
UN Number 1759
Formula Weight 212.00
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Cyanomethyl)piperidinium Tetrafluoroborate